Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Geiger Counter
Large four-digit readout shows counts per minute or continuous counting.
Manufacturer: Science First EN15
Description
- Student-ready, compact and rugged four-digit Geiger counter
- Suitable for the Geiger stand with absorbers (S440974)
- Indicator lamp and audible chirp signal show tube counts
- Internal power supply will operate the 900VDC Geiger tube (S440972) or the smaller 450VDC tube (S440973)
- Front panel has a power switch and a switch to select counts per minute or continuous counting
- Push-button for manual resetting is provided
- Rear panel has connectors for the GM tube and a mute switch for the chirp signal
Order Info
Dimensions: 3.5 x 6.5 x 5.5 in. (8.9 x 16.5 x 14cm).
Specifications
Geiger Counter | |
Color-coded 4mm banana jacks | |
1 min. |
900 or 450VDC | |
LED |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title