Learn More
BioVision IncSupplier Diversity Partner Gel-FAST™ Gel Staining/Destaining Kit: Gel-FAST™ 20 Minutes Gel Staining/Destaining Kit: Fast and Sensitive method for SDS-PAGE gel staining and destaining. Stains 40 mini gels. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K90140
SDS-PAGE is one of the most powerful and commonly used techniques in molecular biology. However, traditional protein gel staining and destaining methods take several hours to finish and leaves a high blue background on the gel reducing the detection sensitivity. BioVision’s Gel-FASTTM 20 Minutes Gel Staining/Destaining Kit provides a fast and sensitive method for SDS-PAGE gel staining and destaining. The procedure requires only 5 minutes for gel staining and 15 minutes for destaining. A total of 20 minutes allow visualization of as little as 5 ng of protein on a water-clear background. The kit contains sufficient solutions to stain and destain up to 40 pieces of protein mini gels. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9968