Learn More
GE Healthcare Gel Processing Magnet Available on GSA/VA Contract for Federal Government customers only.
Prevents gels from sliding over one another.
Manufacturer:  GE Healthcare 80634220
Catalog No. 45-002-440
Add to cart
Description
Description
  • When placed on gel staining tray, magnets prevent gels from sliding over one another
  • Prevents gel from coming in contact with splash guard
  • When placed on corners of gels with plastic backing, magnet helps keep gels submerged
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Prevent gels from sliding over one another, prevents gel from coming in contact with splash guard, keep gels submerged
4/Pk.
Gel Processing Magnet