Learn More
GE Healthcare Gel Staining Tray Packs Available on GSA/VA Contract for Federal Government customers only.
For use with Hoefer Processor Plus fluid delivery system.
Manufacturer:  GE Healthcare 80644518
Catalog No. 45-002-443
Add to cart
Description
Description
  • Pack includes gel staining tray base, PTFE-coated tray, lid
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
gel staining tray base, PTFE-coated tray, lid
29 x 35 cm
Hoefer Processor Plus Fluid Delivery System
Gel Staining Tray Pack