Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Gelatin Blocking Buffer, 1% in PBS
$67.00 - $114.50
Specifications
Concentration | 1% in PBS |
---|---|
Storage Requirements | -30°C to -10°C |
Format | Liquid |
Color | Colorless |
Product Type | Blocking Buffer |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AAJ62755AP
|
Alfa Aesar™
J62755AP |
500 mL |
Each for $67.00
Order in Bulk
Request bulk or custom formats |
|
AAJ62755K2
|
Alfa Aesar™
J62755K2 |
1 L |
Each for $114.50
Order in Bulk
Request bulk or custom formats |
|
Specifications
1% in PBS | |
Liquid | |
Blocking Buffer |
-30°C to -10°C | |
Colorless |
Safety and Handling
Recommended Storage : Store at -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title