Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gelatin, Type B, Powder, N.F., J.T.Baker™

Click to view available options
Quantity:
500g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

9000-70-8
Quantity 500g
CAS 9000-70-8
Physical Form Powder
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.20kg
Chemical Name or Material Gelatin, Type B
Grade NF