Gelatin, Type B, Powder, N.F., J.T.Baker™

Manufacturer: J.T.Baker 212405

Packaging Poly Bottle
Quantity 2.5kg

 View more versions of this product

Catalog No. 02-003-126

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 9000-70-8
    Chemical Name or Material Gelatin, Type B
    Grade NF

    Specifications

    Physical Form Powder
    Quantity 2.5kg
    CAS 9000-70-8
    Packaging Poly Bottle
    Specific Gravity 1L = 1.20kg
    Chemical Name or Material Gelatin, Type B
    Grade NF

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit