Learn More
Gelsolin/GSN Mouse anti-Human, Janelia Fluor 646, Clone: 20, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP105161JF646
Catalog No. NBP15161646
Add to cart
Description
Description
Gelsolin/GSN Monoclonal antibody specifically detects Gelsolin/GSN in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
Gelsolin/GSN
20
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
GSN
Gelsolin purified from human plasma
Protein A or G purified
Apoptosis, Cancer, Cytoskeleton Markers, Tumor Suppressors
Primary
2934
Human
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Actin-depolymerizing factor, ADF, AGEL, Brevin, DKFZp313L0718, gelsolin, gelsolin (amyloidosis, Finnish type)
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Gelsolin (human)