Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gelsolin Mouse, Unlabeled, Clone: 2, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610413

 View more versions of this product

Catalog No. BDB610413


Add to cart

Description

Description

Gelsolin was identified as a result of its ability to sever actin filaments in a Ca2+-dependent manner. The gene for gelsolin encodes an 83 kDa protein that migrates as a 93 kDa polypeptide in SDS-gels. The N-terminal domain contains the calcium-independent actin-severin site, whereas the calcium-dependent site is located in the C-terminal portion of the protein. It exhibits significant homology with villin, another calcium-regulated actin filament severing protein. Gelsolin can be found intracellularly, as well as in a secreted form. However, both forms are encoded by the same gene.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications

Specifications

Gelsolin
2
Unconjugated
Affinity Purified
Human Gelsolin aa. 592-768
150μg
Cell Biology
Primary
Canine, Human, Murine, Rabbit, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.