Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Geltrex™ LDEV-Free Reduced Growth Factor Basement Membrane Matrix Non-distribution product as customer accommodation.

Geltrex™ LDEV-Free Reduced Growth Factor Basement Membrane Matrix

$49.11 - $218.00

Specifications

Endotoxin Concentration Low
For Use With (Application) 3D Cell culture (extracellular matrices), mammalian cell culture, primary cell culture, stem cell culture
Formulation Liquid
Packaging 1 Each
Purity Cell culture grade
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
A1413201 Non-distribution product as customer accommodation. View Documents Gibco™
A1413201
1mL Each for $49.11
Add to cart
 
A1413202 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only. View Documents Gibco™
A1413202
5mL Each for $218.00
Add to cart
 
Description

Description

Geltrex™ LDEV-Free Reduced Growth Factor Basement Membrane Matrix is a soluble form of basement membrane extracted from murine Engelbreth-Holm-Swarm (EHS) tumors and is free of viruses, including lactose dehydrogenase elevating virus (LDEV), sometimes found in this type of preparation. The major components of Geltrex™ matrix include laminin, collagen IV, entactin, and heparin sulfate proteoglycans. It is ideal for the promotion and maintenance of differentiated phenotypes in a variety of cell cultures including primary epithelial cells, endothelial cells, smooth muscle cells, and human induced pluripotent stem cells (iPSC). Geltrex™ matrix offers, on average, a 2-fold reduction in several growth factors. As a result, analysis of specific growth factor effects in applications requiring basement membrane extract can be carried out in a controlled and reproducible format.

Key Features of Geltrex™ LDEV-Free Reduced Growth Factor Basement Membrane Matrix:

  • Free of lactose dehydrogenase elevating virus (LDEV) making it ideal for all types of cell culture and mouse in vivo applications
  • Consistent protein concentration lot-to-lot helps reduce the need to screen lots
  • Protocols for various applications, including tube formation assays, are available

    Gain Reliability and Consistency in Your hESC Studies
    Recent advances in processing EHS tumor cells have resulted in virus free basement membrane extracts. Every lot is tested by RT-PCR methods so you can now have confidence that every lot of Geltrex™ is virus free. Additionally, our consistent manufacturing process helps ensure a protein concentration with very little variation from lot-to-lot, averaging 15 mg/ml so you can have confidence in every purchase.

    Investigate Angiogenesis with Validated Gibco™ Protocols
    There are many applications for Geltrex™ LDEV-Free reduced growth factor basement membrane matrix, each requiring different thicknesses and concentrations. In general, a protein concentration of >5 mg/ml is used for differentiation studies of primary cells. Geltrex™ LDEV-Free reduced growth factor basement membrane matrix has been used to develop tube formation assay protocols using Gibco™ primary cells and specialty media. See this and other angiogenesis assay protocols

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

  • Specifications

    Specifications

    Low
    Liquid
    Cell culture grade
    –20 to –80°C
    Tube(s)
    Sterile
    3D Cell culture (extracellular matrices), mammalian cell culture, primary cell culture, stem cell culture
    1 Each
    For research use only. Not for use in diagnostic procedures
    –20 to –80°C
    Geltrex™ LDEV-free Reduced Growth Factor
    8

    For Research Use Only. Not for use in diagnostic procedures.

    Provide Content Correction

    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit