Learn More
AccuTec Blades GEM™ 3-Facet Single Edge Blades
AccuTec Blades GEM™ 3-Facet Single Edge Blades provide a product solution for any need
$3.59 - $30400.00
Specifications
Back Style Aluminum
For Use With (Application) Cutting Catheter Tubing, Scraping, Manufacturing, Industrial and Medical Applications
Material Stainless Steel
Sterility Non-sterile
Type Single Edge
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
17-989-124
View Documents
AccuTec Blades
620174
10/Dispenser, 200 Dispensers/Cs.
Each for $3.59
Case of 200 EA for $575.00
Add to cart
17-989-122
View Documents
AccuTec Blades
620094
125,400/Cs.
Case for $30,400.00
Add to cart
Description
Description
  • Used in medical device manufacturing and various applications
  • Different packaging available for desired use
  • Aluminum backed
Specifications
Specifications
Aluminum
Stainless Steel
Single Edge
Cutting Catheter Tubing, Scraping, Manufacturing, Industrial and Medical Applications
Non-sterile