Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gemfibrozil, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Poly Pail
25812-30-0
C15H22O3
250.34
HEMJJKBWTPKOJG-UHFFFAOYSA-N
5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoic acid
CC1=CC=C(C)C(OCCCC(C)(C)C(O)=O)=C1
25812-30-0
C15H22O3
250.34
HEMJJKBWTPKOJG-UHFFFAOYSA-N
5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoic acid
CC1=CC=C(C)C(OCCCC(C)(C)C(O)=O)=C1

G1076, 25812-30-0, C15H22O3

Spectrum™ Gemfibrozil, USP is used to reduce cholesterol and triglycerides (fatty acids) in the blood. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 1 kg
CAS Max % 1
Percent Purity 98 to 102%