Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gemin 3 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen Gemin 3
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry
Applications Western Blot, Immunohistochemistry
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125752
View Documents
Novus Biologicals
NBP31042525UL
25 μg
Each for $106.19
Add to cart
 
NB125751
View Documents
Novus Biologicals
NBP310425100UL
100 μg
Each for $352.89
Add to cart
 
Description

Description

Gemin 3 Polyclonal antibody specifically detects Gemin 3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

Gemin 3
Western Blot, Immunohistochemistry
Purified
RUO
Mouse
Component of gems 3, DEAD (Asp-Glu-Ala-Asp) box polypeptide 20, DEAD box protein 20, DEAD box protein DP 103, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 20, 103kD, DKFZp434H052, DP103DEAD-box protein DP103, EC 3.6.4.13, Gemin-3, GEMIN3gemin-3, probable ATP-dependent RNA helicase DDX20, SMN-interacting protein
The immunogen is a synthetic peptide directed towards the C-terminal region of mouse Gemin 3 (NP_059093). Peptide sequence RRPIPARSRLVMLPKVETEAPGLVRSHGEQGQMPENMQVSQFKMVNYSYD
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml, Immunohistochemistry
Unconjugated
Rabbit
Neuroscience
PBS buffer, 2% sucrose
11218
Polyclonal
Affinity purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit