Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Gemin 6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100251
Description
Antibody detects endogenous levels of total GEMIN6.
GEMIN6 is part of a large macromolecular complex, localized to both the cytoplasm and the nucleus, that plays a role in the cytoplasmic assembly of small nuclear ribonucleoproteins (snRNPs). Other members of this complex include SMN (MIM 600354), GEMIN2 (SIP1; MIM 602595), GEMIN3 (DDX20; MIM 606168), GEMIN4 (MIM 606969), and GEMIN5 (MIM 607005).
Specifications
Gemin 6 | |
Polyclonal | |
Unconjugated | |
GEMIN6 | |
2610019B15Rik; 2810470M17Rik; FLJ23459; gem (nuclear organelle) associated protein 6; gem nuclear organelle associated protein 6; gem-associated protein 6; gemin 6; Gemin6; gemin-6; SIP2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
362688, 67242, 79833 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8WXD5, Q9CX53 | |
GEMIN6 | |
A synthesized peptide derived from human GEMIN6(Accession Q8WXD5), corresponding to amino acid residues I115-A165. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction