Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GEMIN2 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177177MFV450
Description
GEMIN2 Polyclonal antibody specifically detects GEMIN2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
GEMIN2 | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
Component of gems 2, Gemin-2, GEMIN2survival of motor neuron protein-interacting protein 1, SIP1-delta, SMN interacting protein 1-delta, SMN-interacting protein 1, survival of motor neuron protein interacting protein 1 | |
Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human SIP1. The immunogen is located within the last 50 amino acids of SIP1. Amino Acid Squence: VSRYFDQRDLADEPS | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
8487 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunocytochemistry | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction