Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GEMIN2 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI4C11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271374AF405

Catalog No. N271374A405


Add to cart

Description

Description

GEMIN2 Monoclonal antibody specifically detects GEMIN2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

GEMIN2
OTI4C11
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
GEMIN2
Full length recombinant protein of human GEMIN2 (NP_003607) produced in E.coli.
Immunogen affinity purified
Core ESC Like Genes, Stem Cell Markers
Primary
8487.0
Immunohistochemistry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
Component of gems 2, Gemin-2, GEMIN2survival of motor neuron protein-interacting protein 1, SIP1-delta, SMN interacting protein 1-delta, SMN-interacting protein 1, survival of motor neuron protein interacting protein 1
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit