Learn More
GEMIN8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA589375
Description
Immunogen sequence: MAAVKASTSK ATRPWYSHPV YARYWQHYHQ AMAWMQSHHN AYRKAVESCF NLPWYLPSAL LPQSSYDNEA AYPQSFYDHH VAWQDYPCSS SHFRRSGQHP RYSSRIQAST KEDQALSKEE EMETESDAEV ECDLSNMEIT EELRQYFAET ERHREERRRQ QQLDAERLDS YVNADHDLYC NTRRSVEAPT ERPGERRQAE MKRLYGDSAA KIQAMEAAVQ LSFDKHCDRK QPKYWPVIPL KF; Positive Samples: A-549, HT-29, HepG2; Cellular Location: Cytoplasm, Nucleus, gem
The SMN complex plays a catalyst role in the assembly of small nuclear ribonucleoproteins (snRNPs), the building blocks of the spliceosome. Thereby, plays an important role in the splicing of cellular pre-mRNAs. Most spliceosomal snRNPs contain a common set of Sm proteins SNRPB, SNRPD1, SNRPD2, SNRPD3, SNRPE, SNRPF and SNRPG that assemble in a heptameric protein ring on the Sm site of the small nuclear RNA to form the core snRNP. In the cytosol, the Sm proteins SNRPD1, SNRPD2, SNRPE, SNRPF and SNRPG are trapped in an inactive 6S pICln-Sm complex by the chaperone CLNS1A that controls the assembly of the core snRNP. Dissociation by the SMN complex of CLNS1A from the trapped Sm proteins and their transfer to an SMN-Sm complex triggers the assembly of core snRNPs and their transport to the nucleus.Specifications
GEMIN8 | |
Polyclonal | |
Unconjugated | |
GEMIN8 | |
GEMIN8 | |
Recombit fusion protein containing a sequence corresponding to amino acids 1-242 of human GEMIN8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
3.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NWZ8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54960 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.