Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Geminin, Mouse anti-Human, Clone: CPTC-GMMN-1, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP27974320UG
Description
Geminin Monoclonal specifically detects Geminin in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.Specifications
Geminin | |
Monoclonal | |
0.2 mg/mL | |
Immunohistochemistry 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
O75496 | |
GMNN | |
Recombinant human full-length Geminin | |
20 μg | |
Breast Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Nuclear Receptors Coactivators and Corepressors, Stem Cell Markers | |
51053 | |
Store at 4°C. | |
IgG1 κ |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array | |
CPTC-GMMN-1 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Gem, geminin, geminin, DNA replication inhibitor, RP3-369A17.3 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction