Learn More
Gemtuzumab, Mouse anti-Many, Alexa Fluor 350, Clone: 975155, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB96171U100UG
Catalog No. FAB96171U10
Add to cart
This item is not returnable. View return policy
Description
Description
Gemtuzumab Monoclonal antibody specifically detects Gemtuzumab in Multi-Species Samples. It is validated for Flow Cytometry.
Specifications
Specifications
Gemtuzumab
975155
Flow Cytometry 0.25-1 ug/10^6 cells
Purified
Full length Gemtuzumab
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Detects Gemtuzumab in Western blots and direct ELISAs. No cross-reactivity with Cetuximab, Ibritumomab, Rituximab, Adalimumab, Tositumomab, Brentuximab, Trastuzumab, Solanezumab, Teprotumumab, and Aducanumab is observed.
Flow Cytometry
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Mouse
IgG2a
100 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Many