Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gemtuzumab, Mouse anti-Many, Alexa Fluor 488, Clone: 975155, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB96171G100UG
This item is not returnable.
View return policy
Description
Gemtuzumab Monoclonal specifically detects Gemtuzumab in Multi-Species samples. It is validated for Flow Cytometry.Specifications
Gemtuzumab | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Full length Gemtuzumab | |
100 μg | |
Primary | |
Many | |
Purified |
Flow Cytometry | |
975155 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Detects Gemtuzumab in Western blots and direct ELISAs. No cross-reactivity with Cetuximab, Ibritumomab, Rituximab, Adalimumab, Tositumomab, Brentuximab, Trastuzumab, Solanezumab, Teprotumumab, and Aducanumab is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title