Learn More
BioTek™ Gen5™ Data Collection and Analysis Software PROMO
Compatible with BioTek microplate readers. BioTek Gen5 Data Collection and Analysis Software has a contemporary interface which provides easy access to all major functions of the software including reading parameters, plate layout, data reduction, online help, wizards, tutorials and sample protocols.
Manufacturer:  BioTek™ GEN5PM
Catalog No. 11-120-510PM
Add to cart
This item is not returnable. View return policy
Description
Description
  • Quick launch buttons on the welcome screen provide links to recently used files, pre-programmed protocols and other key functions such as Quick Read and System Test
  • StepWise™ tools allow limitless combinations of procedural steps and data reduction criteria for a wide variety of applications in a logical and clearly defined sequence that is easily viewed on the screen
  • Analyze data on a single plate or across multiple plates in an experiment
  • Report and PowerExport™ builders are fast and flexible output tools to produce publication-ready reports using data matrices, statistics tables, protocols and plate information
  • For compliance to 21 CFR Part 11, Gen5 Secure provides system, data and protocol protection
  • Multiple user definitions and permission levels make it easy to comply with laboratory policies
System Requirements
  • Windows™ XP or Windows 2000 (Professional Editions)
  • Pentium™III-Class PC (or compatible) processor (500MHz or higher)
  • 512 MB RAM or higher
  • 2 GB hard drive space or higher
  • XGA resolution at 1024 x 768 or higher
  • CD-ROM readable drive
  • Keyboard
  • Mouse
  • Microsoft™ Internet Explorer™ v 5.0 or higher (for online help)
  • Microsoft™ Excel™ 2000 or later (for PowerExport feature)
Specifications
Specifications
Reading parameters, plate layout, data reduction, online help, wizards, tutorials and sample protocols
Gen5 Data Analysis Software
Data Collection and Analysis Software
Microplate readers
Documents
Promotions
Promotions
  • BioTek InstrumentsSupplier Diversity Partner
    Get 50% off BioTek Gen5 and other Liquid Handling Control Software.
    06-30-2019