Learn More
Mac Mod AnalyticalSupplier Diversity Partner GENE 4 C18 GRD. 3.0 - 4.6MM 1 DFS Item
Manufacturer:  Mac Mod AnalyticalSupplier Diversity Partner 5169650N
Genesis 4um C18 Guard (for 3.0 - 4.6mm id columns) 3pk Note: All Grace Analytical and Semi-Preparative HPLC Columns brands including Vydac®, Alltima®, Alltima® HP, Prevail™, Apollo™, Allsep®, Apex™ and Genesis™ analytical HPLC column ranges are now available in the US from MAC-MOD Analytical via Fisher Scientific - Hichrom Limited recently obtained worldwide manufacturing rights to these brands and is offering them in the US through MAC-MOD Analytical. These columns are made with the same exact manufacturing protocols and to the identical specifications previously used by Grace. Category: Chromatography Columns and Supplies GRACE Part Number: 5169650 Alternative Part Number: 5169650
Catalog No. 50-112-0093