Learn More
Invitrogen™ GeneArt™ High-Order Vector Conversion Cassette Non-distribution product as customer accommodation.
A ready-to-use, 3.8kb linear BAC⁄YAC cassette, containing yeast replication and selection elements that can make any E.coli vector compatible with the GeneArt™ High-Order Genetic Assembly System
Manufacturer:  Invitrogen™ A13291
Catalog No. A13291
Add to cart
Description
Description
The GeneArt™ High-Order Vector Conversion Cassette is a ready-to-use, 3.8 Kb linear BAC⁄YAC cassette, containing yeast replication and selection elements that can make any E.coli vector compatible with the GeneArt™ High-Order Genetic Assembly System (cat# A13285 & A13286). Creating your compatible vector is as simple as blunt cloning the cassette into the vector’s multiple cloning site. This product only includes the DNA cassette.
Some of the key features and elements of the Conversion Cassette include:
  • TRP1 to allow selection of yeast transformants in medium lacking tryptophan
  • URA3 as an optional counter selectable marker in yeast
  • ARS⁄CEN origin to allow stable maintenance of the construct in yeast
  • Chloramphenicol resistance for selection in E.coli
  • 6 restriction enzyme sites, including 2 endonuclease homing sites, designed for rare-cutters to simplify the linearization of the final construct
If you prefer to use a commercial ready-to-use vector, please consider the GeneArt™ pYES1L Vector with Sapphire Technology™ (cat# A13287). The vector comes linearized and has been verified to clone up to 110 Kb of DNA fragments.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
  • Structure: Linear
  • Cloning Method: Seamless Cloning
  • Replication Origin: ARS⁄CEN
  • Selection Marker (Eukaryotic): TRP1, URA3
  • Antibiotic Resistance (Bacterial): Chloramphenicol (CmR)
  • Shipping Condition: Dry Ice
The product contains one vial of linear conversion cassette at a concentration of 100ng⁄ml. Each vial contains enough DNA to convert 10 different E.coli vectors.
10 reactions
Store the product at -20°C. The cassette is guaranteed stable for 6 months when properly stored.