Learn More
Invitrogen™ GeneArt™ Type IIs Assembly Kit Non-distribution product as customer accommodation.
Provides seamless cloning and assembly of up to 8 DNA fragments by simultaneous cleavage and ligation in a single reaction
Manufacturer:  Invitrogen™ A15918
 View more versions of this product
Catalog No. A15918
Add to cart
Specifications
Specifications
Bbs I
10 reactions
10 kb total (vector plus all inserts); Up to 8 Fragments
Store in ultra-cold freezer (-68° to -85°C).