Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare GeneGel Excel Electrophoresis Kit 12.5%, 0.5 mm, 24-well Available on GSA/VA Contract for Federal Government customers only.

Separation Range: 100 to 1500 bp; Separation Type: Denaturing; For Use With: GenePhor Electrophoresis System

Manufacturer:  GE Healthcare 17600014

Catalog No. 45-002-380


Add to cart

Description

Description

GeneGel Excel is specifically designed for routine analysis using the GenePhor Electrophoresis System. GeneGel Excel kits contain five precast gels and six pairs of buffer strips that allow you to avoid the hazards of acrylamide and the nuisance of making up multiple buffers. The gel and buffer strip combination constitutes a discontinuous buffer system especially designed for DNA separations.
  • 24 sample wells accommodate up to 6µl of sample
  • Buffer strips are color-coded
  • Total set up time is less than 3 minutes
The GenePhor Electrophoresis System has been specifically designed and tested to meet the critical requirements of separating small DNA fragments typically generated by standard PCR techniques that include SSCP, tandem repeat, RAPD and differential display analysis. The complete GenePhor Electrophoresis System is an integrated platform that includes the GenePhor Electrophoresis Unit, EPS 1001 Power Supply, Processor Plus automated gel staining and blot processing unit, and a wide selection of precast polyacrylamide gels and buffer systems. When using GeneGel Excel gels, simply place the gel onto the bed of the GenePhor Electrophoresis unit direct from the packet.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

1 Each
12.5%
100 to 1500 bp
24-well
T = 6%, C = 3%
GenePhor Electrophoresis System
0.5 mm
Denaturing
5 precast GeneGel Excel 12.5/24 gels; 6 Anode & 6 Cathode buffer strips. 1 manual. These Ready-to-run gels
are 12.5% acrylamide, 0.5mm thick.
Electrophoresis Kit
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit