Learn More
Thermo Scientific™ GeneJET DNA Purification Kits Promotion
Designed for rapid and efficient purification of high quality genomic DNA from various mammalian cell culture and tissue samples, whole blood, bacteria and yeast.
Manufacturer:  Thermo Scientific™ K0721
 View more versions of this product
Catalog No. FERK0721PM
Add to cart
Specifications
Specifications
Isolated genomic DNA is ideal for use in common molecular biology procedures, including: PCR and qPCR FastDigest and conventional restriction digestion Southern blotting
Proteinase K Solution, RNase A Solution, Digestion Solution, Lysis Solution, Wash Buffer I (concentrated), Wash Buffer II (concentrated), Elution Buffer, GeneJET Purification Columns (pre-assembled with collection tubes), Collection tubes, Detailed Protocol
Fermentas GeneJET DNA Purification Kit
50