Learn More
Thermo Scientific™ GeneJET Plant RNA Purification Mini Kit Promotion
Designed for rapid and efficient purification of high quality total RNA from wide variety of plant species and tissue types
Manufacturer:  Thermo Scientific™ K0801
 View more versions of this product
Catalog No. FERK0801PM
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Fast extraction of high purity RNA suitable for all conventional molecular biology procedures, including: RT-PCR,  RT-qPCR, Northern blotting, Nuclease protection assay
Plant RNA Lysis Solution, Wash Buffer WB 1 (concentrated), Wash Buffer 2 (concentrated), Water, nuclease-free, GeneJET RNA Purification Columns (pre-assembled with collection tubes), Collection tubes (2 mL), Collection tubes (1.5 mL), D
50 preps.