Learn More
Thermo Scientific™ GeneJET™ Plasmid Midiprep Kit PROMO
Designed for large-scale isolation of high quality plasmid DNA from recombinant E.coli cultures.
Manufacturer:  Thermo Scientific™ K0481
 View more versions of this product
Catalog No. FERK0481PM
Add to cart
Description
Description
Highlights
  • Efficient -high yields up to 200 µg of high quality plasmid DNA from 50 mL of bacterial culture
  • High concentration of DNA purified -> 400 ng/µL
  • Convenient -spin column format, centrifuge or vacuum protocols
  • Fast -procedure takes 60 minutes, considerably faster than using gravity flow columns
Specifications
Specifications
400ng/ μL
Resuspension Solution, Lysis Solution, Neutralization Solution, Endotoxin Binding Reagent, Wash Solution I (concentrated), Wash Solution II (concentrated), RNase A Solution, Elution Buffer (10mM Tris-HCl, pH 8.5), GeneJET™ Midi Purification Columns pre-assembled with collection tubes (50 mL) and Collection tubes (50 mL).
GeneJET™ Plasmid Midiprep Kit
FastDigest™ restriction enzyme digestion, automated fluorescent sequencing, PCR, cloning, transformation, in vitro transcription, transfection of robust cell lines.
25 Preps