Learn More
Thermo Scientific™ GeneJET™ RNA Cleanup and Concentration Micro Kit PROMO
Perform rapid and efficient concentration of prepurified RNA samples with this kit.
Manufacturer:  Thermo Scientific™ K0842
 View more versions of this product
Catalog No. FERK0842PM
Add to cart
Description
Description
Highlights
  • Advanced column design -easy to open, load and handle.
  • Elute in 6 to 10 µL volume.
  • 85 to 100% recovery of a high integrity RNA.
  • Exceptional purity - isolate RNA free of enzyme inhibitors such as heparin, EDTA or citrate; DNAse I and other enzymes.
  • Fast and simple -procedure takes less than 5 minutes.
  • Applications The purified RNA can be used in common downstream applications:
  • RT-PCR, RT-qPCR, Northern blotting and other RNA-based analysis.
Specifications
Specifications
GeneJET RNA purification Micro column and collection tube, Binding buffer, Wash buffer 1 (conc.), Wash buffer 2 (conc.), Water (nuclease-free), Collection tubes (1.5 mL)
250 preps
Ambient