Learn More
Thermo Scientific™ GeneJET Whole Blood Genomic DNA Purification Mini Kits Promotion
Perform silica membrane-based genomic DNA purification from fresh, frozen anticoagulated mammalian blood, aged blood, buffy coat, bone marrow and body fluids
Manufacturer:  Thermo Scientific™ K0781
 View more versions of this product
Catalog No. FERK0781PM
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • High yields of high molecular weight genomic DNA
  • 20 minute procedure following cell lysis
  • Spin columns are capped and assembled with collection tubes
  • DNA free of enzyme inhibitors, RNA and proteins, ready to use for all demanding applications, validated in realtime PCR, endpoint PCR, multiplex PCR and restriction enzyme digests
  • Compatible: Suitable for wide variety of samples, like fresh or frozen anticoagulated mammalian blood (treated with EDTA, Citrate or Heparine), aged blood, buffy coat, bone marrow, body fluids
Specifications
Specifications
Isolated genomic DNA is ideal for use in common molecular biology procedures, including PCR and qPCR, FastDigest™ and conventional restriction digestion, Southern blotting
Proteinase K Solution, Lysis Solution, Wash Buffer WB I (concentrated), Wash Buffer II (concentrated), Elution Buffer, GeneJET Purification Columns (pre-assembled with collection tubes), Collection tubes, Detailed Protocol
50 reactions