Learn More
Alpco Ltd GeneProof Cytomegalovirus (CMV) PCR Kit
SDP
Supplier:  Alpco Ltd 60CMV002100
Encompass
The GeneProof Cytomegalovirus (CMV) PCR Kit is precise and fully traceable quantification according to 1st WHO International Standard NIBSC 09/162. For Research Use Only in North America. This kit is sufficient for 100 tests.
Catalog No. NC2271533
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.