Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific General Chemistry Calibrators

Liquid-stable, serum-based calibrator sets contain both Total and Direct Bilirubin values

Manufacturer:  Pointe Scientific M7562CAL

 View more versions of this product

Catalog No. 23-666-371


Add to cart
This item is not returnable. View return policy

Description

Description

•    Chemistry Calibrator (23-666-189): human serum-based (lyophilized)
•    Cholesterol Calibrator (23-666-203): human serum-based (lyophilized) with cholesterol value of approximately 50mg/dL

Specifications

Specifications

CE
2°C to 8°C
Microalbumin Calibrator Set
Calibration of pointe scientific's AutoHDL™ and autoLDL™ Cholesterol reagent set in Serum or Plasma, in-vitro diagnostic use only
Clear
Liquid
Sodium Azide 0.001%
6 x 1 mL
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit