Learn More
Stockwell Scientific General Labware Flange Caps
For resealing glass test tubes and glass culture tubes
Manufacturer:  Stockwell Scientific 8559Y
 View more versions of this product
Catalog No. 03-342-250
Add to cart
Description
Description
  • Can be written on with indelible ink for easy sample identification
  • Prevents spills and evaporation by employing a liquid tight seal
  • Available in various transparent colors
Specifications
Specifications
Flanged
LDPE
1000/bag
Yellow
Flange cap w/liquid-tight seal
12mm