Fisherbrand™ General Protection Disposable SMS Blueberry Lab Coats
Single-use lab coats provide high durability and splash/dust protection
 View more versions of this product
Catalog No. 17-100-865A
  Check Availability
Add to Cart
    Specifications
    No. of Pockets 3
    Packaging Type 10/Pk.; 30/Cs.
    Product Type General SMS protection; Three pockets
    Material SMS (Spunbound Melt-Blown Synthetic)
    Size Small
    Disposable Yes
    Collar Type Standard
    Latex Free Yes
    Closure Type Snap
    Color Blueberry
    Length (English) 35 in.
    Cuff Style Knit
  • Description & Specifications
Related Products
Product Content Feedback