Learn More
MilliporeSigma™ Supelco™ General Purpose Copper Tubing
Cleaned to ASTM B-280 Specifications
Supplier: MilliporeSigma™ Supelco™ 20522
Specifications
Tubing | |
Gas Chromatography (GC) | |
1/4 in. | |
6.35 mm | |
Copper |
Cleaned to ASTM B-280 Specifications | |
0.190 in. | |
4.83 mm | |
50 ft. |