Thermo Scientific Value Lab Upright Freezers, 20.0 cu. ft., 120V

Meet routine laboratory storage requirements with Thermo Scientific™ Value Lab Upright Freezers. Freezers feature digital controls and excellent temperature stability.

Manufacturer: Thermo Scientific 20LFEETSA/DEL

Catalog No. 11-675-951

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Temperature Range (Metric) -12°C to -20°C 
    Dimensions (L x W x H) Exterior 32.13 x 34 x 73.13 in. (81.6 x 86.4 x 185.7 cm)
    Dimensions (D x W x H) Interior 23.8 x 27.8 x 57.8 in. (60.3 x 70.5 x 146.7 cm)
    For Use With (Application) Indoor General Storage Of Non-Critical Samples And Reagents
    Typical Lifetime 10 Years
    Min. Clearance 3” On Sides, 3" At Back, 3" On Top
    Lighting Interior - Yes
    Temperature (Setpoint) Adjustable
    Display Temperature - None
    Certifications/Compliance cCSAus
    Color White
    Control Mechanical Dial Thermostat/Inside, Half Way Up Right Wall
    Warranty 13 Months Parts and Labor
    Current 5 Amps/575 Watts
    Capacity 20.0 cu. ft. / 566.34L,20.9 Cubic Feet/591.82 Liters,20.0 cu. ft. / 566.34L
    Casters Adjustable Leveling Feet
    Defrost Manual,Manual Defrost
    Compressor Bottom Mounted
    Door Count 1(1 compartment)
    Door Style Hinged, Opens left to Right, Non-Reversible
    Exterior Finish Epoxy Coated Steel
    Interior Finish Epoxy Coated Steel
    Insulation Polyurethane Foam,Polyurethane Foam
    Model 20LFEETSA
    Plug Type NEMA 5-15P,Yes/Three prong NEMA 5-15P, 3.048 Meters (10 Feet)
    Shelves 4 fixed Interior shelves, 1 Wire Basket, 6 door shelves,4 Fixed Shelves, 6 Fixed Door Shelves
    Refrigerant R134a,100% CFC Free,R134A,CFCFree,R134A,CFCFree
    Type General Purpose Freezer
    Electrical Requirements 115 V/60 Hz
    Voltage 115V
    Shipping Weight (English) 198 lb.
    Shipping Weight (Metric) 89.8kg
    Doors 1 (1 compartment); Hinged, Opens Left to Right, Non-Reversible
    Hertz 60Hz
    Capacity (Metric) 566.34L
    Capacity (English) 20.0 cu. ft.
    Amperage 5.0A,5.0A
    Width (English) Interior 27 in.
    Width (Metric) Exterior 83.8cm
    Width (Metric) Interior 68.58cm
    Depth (English) Interior 21 in.
    Depth (Metric) Interior 53.34cm
    Height (English) Exterior 67.25 in.
    Height (English) Interior 57.5 in.
    Height (Metric) Exterior 170.8cm
    Length (English) Exterior 31.25 in.
    Length (Metric) Exterior 79.4cm
    Width (English) Exterior 33 in.
    Height (Metric) Interior 146.05cm

    Meet routine laboratory storage requirements with Thermo Scientific™ Value Lab Upright Freezers. Freezers feature digital controls and excellent temperature stability.

    • 20 cu. ft. upright
    • Four fixed wire shelves and six door shelves
    • Manual defrost
    • Enameled steel interior and exterior
    • CFC-free insulation and coolant

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit