Thermo Scientific Value Lab Upright Freezers Available on GSA/VA Contract for Federal Government customers only.

Meet routine laboratory storage requirements with Thermo Scientific™ Value Lab Upright Freezers. Freezers feature digital controls and excellent temperature stability.

Manufacturer: Thermo Scientific 20LFEETSV

 View more versions of this product

Catalog No. 20LFEETSV

  Check Availability
Add to Cart

    Specifications

    Dimensions (L x W x H) Exterior 32.13 x 34 x 73.13 in. (81.6 x 86.4 x 185.7 cm)
    Dimensions (D x W x H) Interior 23.8 x 27.8 x 57.8 in. (60.3 x 70.5 x 146.7 cm)
    Temperature Range (Metric) -12°C to -20°C
    For Use With (Application) Indoor General Storage Of Non-Critical Samples And Reagents
    Typical Lifetime 10 Years
    Min. Clearance 3” On Sides, 3" At Back, 3" On Top
    Lighting Interior - Yes
    Temperature (Setpoint) Adjustable
    Display Temperature - None
    Color White
    Control Mechanical Dial Thermostat/Inside, Half Way Up Right Wall
    Warranty 13 Months Parts and Labor
    Current 5 Amps/1150 Watts
    Casters Adjustable Leveling Feet
    Compressor Bottom Mounted
    Door Count 1(1 compartment)
    Door Style Hinged, Opens Left to Right, Non-Reversible
    Exterior Finish Epoxy Coated Steel
    Interior Finish Epoxy Coated Steel
    Model 20LFEETSV
    Type General Purpose Freezer
    Electrical Requirements 230 V/50 Hz
    Capacity 20.9 Cubic Feet/591.82 Liters,20.0 cu. ft. / 566.34L
    Defrost Manual Defrost
    Shelves 4 Fixed Shelves, 6 Fixed Door Shelves
    Plug Type Yes/Three prong NEMA 5-15P, 3.048 Meters (10 Feet)
    Refrigerant R134a,100% CFC Free,R134A,CFCFree
    Shipping Weight (English) 198 lb.
    Shipping Weight (Metric) 89.8kg
    Voltage 230V
    Doors Single (one compartment); Hinged, Opens Left to Right, Non-Reversible
    Certifications/Compliance cULus
    Amperage 5.0A,5.0A
    Width (English) Interior 27 in.
    Capacity (Metric) 566.34L
    Insulation Polyurethane Foam,Polyurethane Foam
    Hertz 50Hz
    Depth (English) Interior 21 in.
    Height (English) Interior 57.5 in.
    Height (Metric) Exterior 170.8cm
    Height (Metric) Interior 146.05cm
    Length (Metric) Exterior 79.4cm
    Capacity (English) 20.0 cu. ft.
    Width (Metric) Exterior 83.8cm
    Height (English) Exterior 67.25 in.
    Length (English) Exterior 31.25 in.
    Depth (Metric) Interior 53.34cm
    Width (English) Exterior 33 in.
    Width (Metric) Interior 68.58cm

    Meet routine laboratory storage requirements with Thermo Scientific™ Value Lab Upright Freezers. Freezers feature digital controls and excellent temperature stability.

    • 20 cu. ft. upright
    • Four fixed wire shelves and six door shelves
    • Manual defrost
    • Enameled steel interior and exterior
    • CFC-free insulation and coolant

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit