Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Value Lab Upright Freezers Available on GSA/VA Contract for Federal Government customers only.

Meet routine laboratory storage requirements with upright freezers that feature digital controls and excellent temperature stability.

Manufacturer:  Thermo Scientific™ 05LFEETSA

 View more versions of this product

Catalog No. 05LFEETSA


Add to cart
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Description

Description

Meet routine laboratory storage requirements with Thermo Scientific™ Value Lab Upright Freezers. Freezers feature digital controls and excellent temperature stability.

1.6 cu. ft. Model

  • Suitable for -12° to -20°C
  • One fixed interior shelf
  • Key lock
  • UL listed

3.5 cu. ft. Model

  • Suitable for -12° to -20°C
  • Three fixed bins
  • Adjustable thermostat with deep-freeze setting
  • Overtemperature warning with visual alarm
  • Built-in key lock

5 cu. ft. Model

  • Suitable for -12° to -20°C
  • Two fixed shelves
  • Without key lock

Includes: Adjustable shelves or baskets

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

-12°C to -24°C
5 cu. ft.
115 V 60 Hz
60 Hz
NEMA 5-15 P
19 x 18.63 x 29.5 in. (48.2 x 47.32 x 74.9)
32.7 kg
43 kg
R134a, 100% CFC Free
None
White
Indoor General Storage Of Non-Critical Samples And Reagents
1 in. On Sides, 1 in. At Back, 4 in. On Top
Adjustable
05LFEETSA
General Purpose Freezer
Hinged, Opens Left to Right, Reversible
Polyurethane Foam
1 (1 compartment)
2 Fixed Shelves, 3 Door Shelves
cULus
141.6 L
115 V
1.1 A
25.75 x 21.38 x 33.13 in. (84.15 x 54.3 x 65.4)
72 lb.
95 lb.
Temperature Display - None
ABS Plastic
Warranty varies by country, please contact us for details
Enameled Steel
10 Years
Interior - Yes
2 Adjustable Feet In Front, 2 Fixed Pads In Rear
Manual
Mechanical Dial Thermostat/ Inside, Side wall top
Bottom Mounted
Alarm
10
Undercounter
SDS
Documents
Product Certifications
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit