Learn More
Globe Scientific General Purpose Polypropylene Microcentrifuge Tubes
Feature attached leak-resistant flat top snap caps
$32.55 - $347.00
Specifications
Volume (Metric) 2 mL
Graduated Yes
Max. RCF 18000 rpm
Material Polypropylene
Sterility Non-sterile
View More Specs
Products 3
Catalog Number Mfr. No. Cap Style No. per Pack Color Price Quantity & Availability  
Catalog Number Mfr. No. Cap Style No. per Pack Color Price Quantity & Availability  
22-010-088 View Documents Globe Scientific
111552
Flat top cap 1000 Natural Pack of 1000 for $72.00
Add to cart
22-010-091 View Documents Globe Scientific
111568
Snap cap 1000 Natural Pack of 1000 for $32.55
Add to cart
22-010-098 View Documents Globe Scientific
111552A
Flat top cap 5000 Amber Case of 5000 for $347.00
Add to cart
Description
Description
  • Molded with precise graduation marks for accurate measurements
  • Natural tubes have round bottom
  • Amber tubes have conical bottom
Specifications
Specifications
2 mL
18000 rpm
Non-sterile
Yes
Polypropylene
Yes