Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Genes and Probability
Discover the fundamental principles of genetics.
Specifications
For Use With (Application) | Discover the fundamental principles of genetics, monohybrid and dihybrid crosses, laws of dominance, segregation and recombination |
---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S63066
![]() |
Neo/Sci Corporation
201163 |
Genes And Probability Genetic Kit |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Students use game-like exercises to explore gametogenesis and fertilization; monohybrid and dihybrid crosses; laws of dominance, segregation, and recombination; independent assortment and other key elements of this important and relevant topic.
- Allows students to exercise their skills in observation, classification and comparison
- Contains everything needed to perform the lab activity, including game pieces and comprehensive teacher and student guides
- Great value. Save money when you buy this kit with its corresponding CD-ROM (S63394) as a curriculum pack. For more information on the CD-ROM see the Multimedia Section of this catalog.
Order Info
Class size: 40 students.
Specifications
Discover the fundamental principles of genetics, monohybrid and dihybrid crosses, laws of dominance, segregation and recombination |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title