Learn More
Thermo Scientific™ GENESYS™ 10S Vis Spectrophotometer
This product is discontinued. Please see other models in the Thermo Scientific™ GENESYS™ UV-Vis product line.
Manufacturer:  Thermo Scientific™ 840207900
 View more versions of this product
Catalog No. 14-386-519
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This product is discontinued. Please see other models in the Thermo Scientific™ GENESYS™ UV-Vis product line.
This product is discontinued. Please check out the Thermo Scientific™ GENESYS™ UV-Vis Spectrophotometer product line.
Specifications
Specifications
Single Beam
Single cell holder and automated 6-cell changer standard
15.7 in.
GENESYS 10S Visible
In Germany, France, and Spain, use Catalog #840-209500
Graphical with LCD backlight, 9.7 x 7.1 cm (3.8 x 2.8 in)
Selected automatically, 100/240V 50/60Hz
25cm
Sealed membrane with tactile response keys
<0.001 A at 0.0 A<0.002 A at 2.0 APeak-to-peak at 340 nm
0.5% or ±0.005A, whichever is greater up to 2A
Up to 3A at 340nm
Up to 3600nm/min
<0.1%T at 340, and 400nm
±1.0nm
±0.5nm
8.6kg
30cm
USB Type A port for USB memory device (front panel)USB Type B port for optional computer connectivity (rear panel)USB Type A port for external printer (rear panel)
1.0, 2.0, 3.0, 5.0 nm
40cm
Silicon Photodiode
<0.002A/hr after warm-up
9.8 in.
USB cable, USB memory device
Tungsten-Halogen Lamp (1000 hrs.)
Single Beam
-0.1-3.0A; -0.3-125%T; ±9999C
Internal printer:40 column Internal (text and graphics)Included with PNs 840-208000 and 840-209600Optional accessory for PNs 840-207900 and 840-209500External printer support:External printer with USB connection and HP PCL 3.0 or higher printer language support
5nm
100/240V
325 to 1100nm
19 lb.
11.8 in.
Documents
Accessories