Learn More
Thermo Scientific™ GENESYS™ 30 Tungsten-Halogen Lamp, Pre-aligned
Manufacturer:  Thermo Scientific™ 840278700/EMD
Catalog No. S28522
Add to cart
Description
Description
This Tungsten-Halogen pre-aligned lamp is so easy to replace you might even enjoy replacing it!
Pre-aligned, pre-assembled replacement lamp
Customer replaceable - slides into place
Mouse-hole replacement access.  Cover removal not required.
Specifications
Specifications
Pre-aligned, pre-assembled replacement Tungsten- Halogen lamp. Mouse-hole replacement access. Cover removal not required.
GENESYS 30 Visible Spectrophotometer
1 pre-aligned lamp assembly