Learn More
Lab-Aids™ Genetic Equilibrium and Natural Selection Products for Science Education
Students study the principles of genetic equilibrium and natural selection as a process of evolution. Lab-Aids™ Genetic Equilibrium and Natural Selection is a practical simulation to help students understand these concepts.
Manufacturer:  Lab-Aids™ 76
Catalog No. S20971
Add to cart
Includes: Teacher's guide; 30 student worksheets and guides; 15 shakers; 60 alleles, plastic, dark-colored; 40 alleles, plastic, light-colored
Description
Description
Students study the principles of genetic equilibrium and natural selection as a process of evolution using this kit. A simulation is used to develop insights about the genetic process involved when natural selection operates within a population in genetic equilibrium. Students use the data collected in the Hardy-Weinberg Equilibrium to understand better the actual complexity of natural selection. Accommodates unlimited classes, each with 15 groups of 2.
Specifications
Specifications
45 students/15 groups
Teacher's guide; 30 student worksheets and guides; 15 shakers; 60 alleles, plastic, dark-colored; 40 alleles, plastic, light-colored
Kit: Genetic equilibrium and natural selection
Education