Learn More
Genetic Traits Test Strips Products for Science Education
A quick, easy, and completely safe means to analyze commonly tested genetic traits.
Specifications
For Use With (Application) To determine their possible genotypes for the trait and the frequency of tasters and non-tasters in a population, as well as construct a family pedigree showing the heredity of the trait
View More Specs
Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S85286 Products for Science Education
View Documents
Neo/Sci Corporation
992405
Control Taste Strips This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
S85288 Products for Science Education
View Documents
Neo/Sci Corporation
605134
Sodium Benzoate Test Strips This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Simple taste tests allow students to determine their possible genotypes for the trait and the frequency of tasters and non-tasters in a population, as well as construct a family pedigree showing the heredity of the trait.
Four types available:
  • PTC taste test strips
  • Sodium benzoate taste test strips
  • Thiourea taste test strips
  • Control taste test strips
Specifications
Specifications
To determine their possible genotypes for the trait and the frequency of tasters and non-tasters in a population, as well as construct a family pedigree showing the heredity of the trait