Learn More
Genlantis™ gWiz™ Vectors
High-level transient expression in mammalian cells
Supplier: Genlantis™ AMS.P040400
Description
- High levels of transient gene expression
- Suitable for in vitro and in vivo gene expression applications
- High plasmid yield in E. coli
- Expression ready with the following popular reporter genes: galactosidase, green fluorescent protein (GFP), luciferase, chloramphenicol acetyltransferase (CAT), secreted alkaline phosphatase (SEAP)
- Systematically engineered to produce the highest levels of reporter gene expression in a wide variety of mammalian cells and tissues
Specifications
GenePORTER 2 | |
High | |
25μL TE Buffer (10mM Tris HCl, pH 7.5, 1mM EDTA) | |
25 μg | |
Produce the highest levels of transgene expression in a wide range of mammalian cells and tissues |
CMV | |
Green Fluorescent Protein Vector | |
-20°C | |
Expression vector |