Learn More
Gemini Bio ProductsSupplier Diversity Partner GENTAMICIN SOLN 10MG/ML 6X20ML DFS Item
Manufacturer:  Gemini Bio ProductsSupplier Diversity Partner 400107
Gentamicin sulfate kills Gram-negative and Gram-positive bacteria, as well as mycoplasmas. It works by binding to microbial ribosomes and preventing the initiation of protein synthesis. Recommended dosing in media is 5 mL/L.
Catalog No. 50-753-3029