Learn More
Affymetrix/USB™ Gentamicin Sulfate, Ultrapure
Inhibits normal protein synthesis
Manufacturer:  Affymetrix/USB™ J1605114
 View more versions of this product
Catalog No. AAJ1605114
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
  • Bactericidal
  • Stable
  • Autoclavable
  • Effective when combined with antibiotic that interferes with cell wall synthesis
Specifications
Specifications
Bactericidal
Powder
3.5 to 5.5
Ultrapure
Requires no refrigeration
≥590μg/mg (Dry basis)
Plastic bottle
Antibiotic
25g