Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Affymetrix/USB™ Gentamicin Sulfate, Ultrapure

Inhibits normal protein synthesis

$78.20 - $637.50

Specifications

Product Type Antibiotic
Antibiotic Spectrum Bactericidal
Concentration ≥590μg/mg (Dry basis)
Format Powder
pH 3.5 to 5.5
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AAJ1605103 View Documents Affymetrix/USB™
J1605103
1g Plastic bottle Each for $78.20
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ1605106 View Documents Affymetrix/USB™
J1605106
5g Plastic bottle Each for $248.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ1605114 View Documents Affymetrix/USB™
J1605114
25g Plastic bottle Each for $637.50
Order in Bulk

Request bulk or custom formats

Add to cart
 
Description

Description

  • Bactericidal
  • Stable
  • Autoclavable
  • Effective when combined with antibiotic that interferes with cell wall synthesis
Specifications

Specifications

Antibiotic
≥590μg/mg (Dry basis)
3.5 to 5.5
Requires no refrigeration
Bactericidal
Powder
Ultrapure
Safety and Handling

Safety and Handling

EINECSNumber : 215-778-9

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit