Gentian Violet, 1% (w/v) Alcoholic Solution, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 323216

Packaging Poly natural
Quantity 500mL

 View more versions of this product

Catalog No. 3232-16

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 64-17-5,548-62-9,67-56-1 (Methanol (Methyl Alcohol))
    CAS Min % 97.0,1.21
    CAS Max % 100.0,1.26
    Physical Form Liquid
    Chemical Name or Material Gentian Violet

    Specifications

    Physical Form Liquid
    Color Clear, dark violet
    Quantity 500mL
    Concentration or Composition Notes 1% (w/v)
    Solvent or Matrix Alcoholic Solution
    Packaging Poly natural
    CAS 64-17-5,548-62-9,67-56-1 (Methanol (Methyl Alcohol))
    CAS Min % 97.0,1.21
    CAS Max % 100.0,1.26
    Chemical Name or Material Gentian Violet

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit