Learn More
GeoSafari™ Motorized Solar System and Planetarium
Create the solar and star systems in your classroom.
Supplier: Educational Insights 8702
Alert:
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Includes: Central tower, eight planet orbs plus one dwarf planet orb with identifying planetary symbols, nine metal rods, sun sphere, northern hemisphere star dome, krypton bulb
Description
- The “sun” shines light onto the nine planets as they orbit around it
- The central tower and planetary support rods are black so they “disappear” in a darkened room
- Replace the solar dome with the star dome to create a spectacular planetarium sky complete with major constellation names
- Base features a special 1000-hour krypton light bulb and planetary longitude marks; marks help user set up planets in correct positions
Order Info
Grades 1+. Powered by four C batteries (S43922H) or by an optional AC adapter (S96077).
Specifications
Adapter | |
Each |